Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloride Standard, 1000ppm, Ricca Chemical
SDP

Catalog No. SC023100500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Poly natural; 500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SC023100500 Poly natural; 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SC023100500 Supplier Ricca Chemical Company Supplier No. SC023100500A

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Natural Poly Bottle
CAS 7647-14-5, 7732-18-5
Concentration or Composition Notes 1000ppm
CAS Min % 98.0, 0.16
CAS Max % 100.0, 0.17
Equivalence 1 mL = 1 mg Cl
Physical Form Liquid
Molecular Formula ClNa
MDL Number MFCD00003477
Color Colorless
Boiling Point 100°C
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Quantity 500 mL
PubChem CID 5234
ChEBI CHEBI:26710
Chemical Name or Material Chloride Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.