Learn More
Aqua Solutions Chloride Std. 100.0 ppm (1ml = 0.1mg Cl), NIST Traceable (100ML)
SDP
Catalog No. NC9634143
Encompass
Chloride Std. 100.0 ppm (1ml = 0.1mg Cl), NIST Traceable (100ML)
Catalog No. NC9634143 Supplier Aqua Solutions Supplier No. AM084
Chloride Std. 100.0 ppm (1ml = 0.1mg Cl), NIST Traceable (100ML)