Learn More
Aqua Solutions CHLORIDE STD 1000 PPM
SDP
Catalog No. NC9070019
Encompass_Preferred
Chloride Standard 1000ppm w/w Traceable to N.I.S.T., Components: Water, Deionized ASTM Type II, CAS No.: 7732-18-5, Sodium Chloride, CAS No.: 7647-14-5, <0.5%, Melting Point: ~0degreeC, Specific Gravity: 1, Boiling Point: ~100degreeC
Catalog No. NC9070019 Supplier Aqua Solutions Supplier No. 1764500ML
Chloride Standard 1000ppm w/w Traceable to N.I.S.T., Components: Water, Deionized ASTM Type II, CAS No.: 7732-18-5, Sodium Chloride, CAS No.: 7647-14-5, <0.5%, Melting Point: ~0degreeC, Specific Gravity: 1, Boiling Point: ~100degreeC