Learn More
ERA WATERS CORP CHLORIDE STD 1000MG/L 500ML
SDP
Catalog No. NC9738459
Encompass
Chloride - 1000 mg/l, 500 ml One 500 mL standard containing Chloride at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.
Catalog No. NC9738459 Supplier ERA WATERS CORP Supplier No. 988
Chloride - 1000 mg/l, 500 ml One 500 mL standard containing Chloride at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.