Learn More
SPEX CERTIPREP LLC 3P CHLORIDE STD 100PPM/100ML
Catalog No. NC9799482
Encompass_Preferred
(HPS by Zeptometrix) 100 ug/mL Chloride in H2O; Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC9799482 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICCL100
(HPS by Zeptometrix) 100 ug/mL Chloride in H2O; Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.