Learn More
Hach Company Chloride Standard Solution, 100 mg/L, 1 L
Supplier:  Hach Company 2370853
Encompass
ISE Standard Solution. Chloride, 100 mg/L ±1 mg/L as Cl- (NIST). 1 L bottle
Catalog No. 50-199-5880