Learn More
Hach Company Chloride Standard Solution, 1000 mg/L, 500 mL
Supplier:  Hach Company 18349
Encompass
ISE Standard Solution. Chloride, 1000 mg/L ±10 mg/L as Cl- (NIST). 500 mL bottle
Catalog No. 50-199-4190