Learn More
Cayman Chemical Chlorido[N,N'-disalicylidene-1,2-phenylenediamine]iron(III), Purity: ≥95%, CAS: 39916-28-4, 100 mg
SDP
Catalog No. 502937213
Encompass_Preferred
An inducer of ferroptosis; induces ferroptosis, but not apoptosis or necroptosis, in NB1 cancer cells at 3 µM; inhibits proliferation of BJAB, NALM-6, Jurkat, MelHO, and MCF-7 cancer cells (IC50s = 0.07, 2.5, 1.5, 3 and 5 µM), as well as daunorubicine- and vincristine-resistant NALM-6 cells at 0.04-0.125 µM
Catalog No. 50-293-7213 Supplier Cayman Chemical Supplier No. 28788100 MG
An inducer of ferroptosis; induces ferroptosis, but not apoptosis or necroptosis, in NB1 cancer cells at 3 µM; inhibits proliferation of BJAB, NALM-6, Jurkat, MelHO, and MCF-7 cancer cells (IC50s = 0.07, 2.5, 1.5, 3 and 5 µM), as well as daunorubicine- and vincristine-resistant NALM-6 cells at 0.04-0.125 µM