Learn More
Apexbio Technology LLC Chlorin e6(Synonyms: Ce6, Chlorophyllin e6, Chlorophyll derivative e6, Chlorin-e6, Chlorin e-6), 50mg, CAS: 19660-77-6.
SDP
Catalog No. 5000060702
Encompass_Preferred
Chlorin e6 (Ce6 CAS 19660-77-6) is a second-generation photosensitizer utilized in photodynamic therapy (PDT) research with demonstrated anticancer activity when activated by laser irradiation Upon irradiation Ce6 generates reactive oxygen species mediating cytotoxic effects and cellular apoptosis In mouse models intravenous administration of Ce6 (2 5 10 mg/kg) combined with irradiation (50 200 J/cm ) resulted in complete elimination of implanted fibrosarcomas In clinical trials for bronchogenic superficial squamous cell carcinoma Ce6-based PDT (40 mg/m 100 J/cm ) produced complete response rates up to 82 9% underscoring its utility for cancer therapy studies
Catalog No. 50-000-60702 Supplier Apexbio Technology LLC Supplier No. B831450
Chlorin e6 (Ce6 CAS 19660-77-6) is a second-generation photosensitizer utilized in photodynamic therapy (PDT) research with demonstrated anticancer activity when activated by laser irradiation Upon irradiation Ce6 generates reactive oxygen species mediating cytotoxic effects and cellular apoptosis In mouse models intravenous administration of Ce6 (2 5 10 mg/kg) combined with irradiation (50 200 J/cm ) resulted in complete elimination of implanted fibrosarcomas In clinical trials for bronchogenic superficial squamous cell carcinoma Ce6-based PDT (40 mg/m 100 J/cm ) produced complete response rates up to 82 9% underscoring its utility for cancer therapy studies