Learn More
Medchemexpress LLC Chlorin e6 trimethyl ester | 35038-32-5 | 99.8% | 638.75 | 100 MG
SDP
Catalog No. 5000349154
Encompass_Preferred
Chlorin e6 trimethyl ester is a methyl pheophorbide-a derivative and a photosensitizer primarily used in photodynamic therapy (PDT). This compound is intended for research purposes only.
  • Methyl pheophorbide-a derivative
  • Functions as a photosensitizer
  • Used in photodynamic therapy (PDT) applications
  • Targets reactive oxygen species (ROS)
  • Relevant to immunology/inflammation, metabolic enzyme/protease, and NF-κB pathways
Catalog No. 50-003-49154 Supplier Medchemexpress LLC Supplier No. HY137475100MG
Chlorin e6 trimethyl ester is a methyl pheophorbide-a derivative and a photosensitizer primarily used in photodynamic therapy (PDT). This compound is intended for research purposes only.
  • Methyl pheophorbide-a derivative
  • Functions as a photosensitizer
  • Used in photodynamic therapy (PDT) applications
  • Targets reactive oxygen species (ROS)
  • Relevant to immunology/inflammation, metabolic enzyme/protease, and NF-κB pathways