Learn More
Medchemexpress LLC Chlorin e6 trimethyl ester | 35038-32-5 | 99.8% | 638.75 | 25 MG
SDP
Catalog No. 5000351861
Encompass_Preferred
Chlorin e6 trimethyl ester is a methyl pheophorbide-a derivative functioning as a photosensitizer for photodynamic therapy (PDT). This solid, brown to black compound exhibits photo cytotoxic activity against human HepG2 cells and is intended for research use only.
  • Used as a photosensitizer
  • Applicable in photodynamic therapy (PDT)
  • Exhibits photo cytotoxic activity
  • Solid appearance with brown to black color
  • Soluble in DMSO
Catalog No. 50-003-51861 Supplier Medchemexpress LLC Supplier No. 25MGHY137475
Chlorin e6 trimethyl ester is a methyl pheophorbide-a derivative functioning as a photosensitizer for photodynamic therapy (PDT). This solid, brown to black compound exhibits photo cytotoxic activity against human HepG2 cells and is intended for research use only.
  • Used as a photosensitizer
  • Applicable in photodynamic therapy (PDT)
  • Exhibits photo cytotoxic activity
  • Solid appearance with brown to black color
  • Soluble in DMSO