Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorin e6 trimethyl ester | 35038-32-5 | 99.8% | 638.75 | 50 MG
SDP

Catalog No. 5000352000
Encompass_Preferred

Chlorin e6 trimethyl ester, a methyl pheophorbide-a derivative, is a photosensitizer that can be used in photodynamic therapy (PDT).

  • Photosensitizer for photodynamic therapy
  • Methyl pheophorbide-a derivative
  • High purity (99.8%)
  • Solid appearance, brown to black color
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 10 mg/mL
  • For research use only

Catalog No. 50-003-52000 Supplier Medchemexpress LLC Supplier No. HY13747550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorin e6 trimethyl ester, a methyl pheophorbide-a derivative, is a photosensitizer that can be used in photodynamic therapy (PDT).

  • Photosensitizer for photodynamic therapy
  • Methyl pheophorbide-a derivative
  • High purity (99.8%)
  • Solid appearance, brown to black color
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 10 mg/mL
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.