Learn More
Medchemexpress LLC Chlorin e6 trimethyl ester | 35038-32-5 | MFCD09838560 | 99.8% | 638.75 | C37H42N4O6 | 5 MG
SDP
Catalog No. 5000222427
Encompass_Preferred
Chlorin e6 trimethyl ester is a methyl pheophorbide-A derivative photosensitizer used in photodynamic therapy research and photophysics studies. It is supplied as a high-purity solid for in-vitro use and formulation, with defined solubility and storage recommendations to preserve stability.
  • High reported purity (99.8%) suitable for research applications.
  • Photosensitizer used in photodynamic therapy and photophysics studies.
  • Soluble in DMSO at about 10 mg/mL with sonication and warming to 60°C; hygroscopic DMSO can affect solubility.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years); in solvent: -80°C (6 months) or -20°C (1 month).
  • Provided as a brown to black solid appropriate for formulation and analytical work.
Catalog No. 50-002-22427 Supplier Medchemexpress LLC Supplier No. HY1374755MG
Chlorin e6 trimethyl ester is a methyl pheophorbide-A derivative photosensitizer used in photodynamic therapy research and photophysics studies. It is supplied as a high-purity solid for in-vitro use and formulation, with defined solubility and storage recommendations to preserve stability.
  • High reported purity (99.8%) suitable for research applications.
  • Photosensitizer used in photodynamic therapy and photophysics studies.
  • Soluble in DMSO at about 10 mg/mL with sonication and warming to 60°C; hygroscopic DMSO can affect solubility.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years); in solvent: -80°C (6 months) or -20°C (1 month).
  • Provided as a brown to black solid appropriate for formulation and analytical work.