Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorinated Herbicides Mixture, 16 Analytes, various concentrations, Ultra Scientific

Catalog No. ushbm5155a1
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USHBM-5155A-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USHBM-5155A-1 Supplier Agilent Technologies Supplier No. HBM5155A1
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Chlorinated Herbicides Mixture
CAS 1634-04-4, 50594-66-6, 25057-89-0, 133-90-4, 94-75-7, 75-99-0
CAS Max % 99.45, 0.01, 0.02, 0.01, 0.02, 0.17
Analyte or Component Names tert-butylmethyl ether (MTBE),acifluorfen,bentazon
Element Tert-butylmethyl Ether, Acifluorfen, Bentazon
Solvent or Matrix MTBE
Quantity 1 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.