Learn More
MSC Zep FS Foam Chlorinated Liquid Detergent, Gallon, 4 cs
Catalog No. 502815745
Encompass
Zep FS Foam Chlorinated Liquid Detergent is formulated for effective cleaning and sanitation in food and beverage facilities.
  • Foaming formula for improved surface contact
  • Contains chlorine for enhanced sanitizing action
  • Suitable for use in food processing environments
  • Liquid formulation for easy application
Catalog No. 50-281-5745 Supplier MSC Supplier No. 17641465
Zep FS Foam Chlorinated Liquid Detergent is formulated for effective cleaning and sanitation in food and beverage facilities.
  • Foaming formula for improved surface contact
  • Contains chlorine for enhanced sanitizing action
  • Suitable for use in food processing environments
  • Liquid formulation for easy application