Learn More
Medchemexpress LLC Chlorindanol | 145-94-8 | 98.0% | 50 MG
SDP
Catalog No. 5000407519
Encompass_Preferred
Chlorindanol (7-Chloro-4-indanol) is a topical antiseptic and sanitizer. It exhibits low systemic toxicity, good skin, eyes, and genital mucosa tolerance, and is nonallergenic. It is rapidly lethal to various microorganisms in vitro, including vegetative bacteria, Trichophyton sp., C. albicans, E. histolytica cysts and trophozoites, T. vaginalis, and spermatozoa.
  • Topical antiseptic or sanitizer.
  • Rapidly lethal to various microorganisms in vitro.
  • Exhibits low systemic toxicity.
  • Good skin, eyes, and genital mucosa tolerance.
  • Nonallergenic.
Catalog No. 50-004-07519 Supplier Medchemexpress LLC Supplier No. HYB099950MG
Add to Cart
Add to Cart
Chlorindanol (7-Chloro-4-indanol) is a topical antiseptic and sanitizer. It exhibits low systemic toxicity, good skin, eyes, and genital mucosa tolerance, and is nonallergenic. It is rapidly lethal to various microorganisms in vitro, including vegetative bacteria, Trichophyton sp., C. albicans, E. histolytica cysts and trophozoites, T. vaginalis, and spermatozoa.
  • Topical antiseptic or sanitizer.
  • Rapidly lethal to various microorganisms in vitro.
  • Exhibits low systemic toxicity.
  • Good skin, eyes, and genital mucosa tolerance.
  • Nonallergenic.