Learn More
Hach Company Chlorine Standard Solution, 50-75 mg/L as Cl2, pk/20 - 2 mL PourRite« Ampules (NIST)
Catalog No. 501994976 Shop All Hach Company Products
Encompass
For accuracy checks in chlorine determinations. Pack of 20 2-mL ampules. Lot-specific concentration displayed on label.
Catalog No. 50-199-4976 Supplier Hach Company Supplier No. 1426820
For accuracy checks in chlorine determinations. Pack of 20 2-mL ampules. Lot-specific concentration displayed on label.