Learn More
Hach Company Chlorine Standard Solution, 50-75 mg/L as Cl2, pk/16 - 10 mL Voluette Ampules (NIST)
Catalog No. 501994975 Shop All Hach Company Products
Encompass
For accuracy checks in chlorine determinations. Pack of 16 10-mL ampules. Lot-specific concentration displayed on label.
Catalog No. 50-199-4975 Supplier Hach Company Supplier No. 1426810
For accuracy checks in chlorine determinations. Pack of 16 10-mL ampules. Lot-specific concentration displayed on label.