Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics Chlorine Activator Solution
SDP

Catalog No. 13299217B
Change view
Click to view available options
Quantity:
6 x 10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-299-217B 6 x 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-217B Supplier CHEMetrics Supplier No. A2500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The pack supplies enough solution to perform at least 200 tests. Used to determine total chlorine.

Specifications

Analytes Chlorine
Format Liquid
Includes (6) 10 mL bottles
Quantity 6 x 10mL
Type Chlorine Activator Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.