Learn More
LaMotte™ Chlorine Bleach Test Kit
SDPSCI_ED
Catalog No. S72400 Shop All LaMotte Company Products
Change view
Click to view available options
Range:
0 to 10%, 0.2% sensitivity
Catalog No. Range
S72400 0 to 10%, 0.2% sensitivity
1 options
Catalog No. S72400 Supplier LaMotte™ Supplier No. 710503
Add to Cart
Add to Cart
Direct Reading Titrator - Test uses a 1.0 mL microburet calibrated to allow direct reading of the test result within a specific range
  • Chlorine Bleach, Iodometric Titration - Test uses an iodine titration to cover the range 0 to 10% chlorine bleach
  • 50 tests at 10% per kit
Specifications
For Use With (Application) Measures for chlorine bleach in industrial water boiling and cooling, water and wastewater
Includes 2 x 30 mL hypochlorite reagent A, 10 g potassium iodide crystals, 60 mL hypochlorite reagent D, test tube with cap, direct reading titrator, 0.5 mL plastic pipet, 1 mL plastic pipet, plastic spoon
No. of Tests 50
Range 0 to 10%, 0.2% sensitivity
Test Method Iodometric Titration
Type Chlorine Bleach Kit