Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Chlorine Dioxide Neutralizer Solution Pack, Use with K-2705
SDP

Catalog No. 13299644
Change view
Click to view available options
Quantity:
6 x 10 mL Bottles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-299-644 6 x 10 mL Bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-644 Supplier CHEMetrics™ by AquaPhoenix Supplier No. A2700
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorine dioxide neutralizer solution accessory pack for compatible test kits. Supplies enough solution to perform at least 200 tests.

  • Includes six 10 mL bottles

Specifications

pH Range 7
Volume (Metric) 0.5 mL
Quantity 6 x 10 mL Bottles
Type Neutralizer Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.