Learn More
Hach Company Chlorine Dioxide Reagent 3, 100 mL
Supplier:  Hach Company 2070242
Encompass
For determination of chlorine dioxide (CIO2) by the chlorophenol red method.
Catalog No. 50-199-5217