Learn More
Grainger Chlorine Dioxide Sensor 0 - 2 ppm PVC
Catalog No. 502768092
Encompass_Preferred
The Hach Chlorine Dioxide Sensor 0 - 10 ppm is an amperometric sensor that measures chlorine dioxide levels in water. It has a measurement range of 0 to 10 ppm chlorine dioxide with an accuracy of ±0.05 ppm. The sensor has a response time of 60 seconds and operates at temperatures between 32-122°F. It features a replaceable membrane cap and requires calibration with chlorine dioxide standards. The sensor connects to a meter or controller and provides continuous monitoring of chlorine dioxide disinfection levels in water treatment applications. It has a 12 month typical electrode life.
Catalog No. 50-276-8092 Supplier Grainger Supplier No. 16Z051
The Hach Chlorine Dioxide Sensor 0 - 10 ppm is an amperometric sensor that measures chlorine dioxide levels in water. It has a measurement range of 0 to 10 ppm chlorine dioxide with an accuracy of ±0.05 ppm. The sensor has a response time of 60 seconds and operates at temperatures between 32-122°F. It features a replaceable membrane cap and requires calibration with chlorine dioxide standards. The sensor connects to a meter or controller and provides continuous monitoring of chlorine dioxide disinfection levels in water treatment applications. It has a 12 month typical electrode life.