Learn More
LaMotte™ Chlorine DPD #3 Instrument Grade Tablets
SDPSCI_ED
Catalog No. S72418 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
S72418 100 Pack
1 options
This item is not returnable. View return policy
Catalog No. S72418 Supplier LaMotte™ Supplier No. 6197AJ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Economical choice for your chlorine testing needs
  • Chemical reagent used to test for total chlorine
  • 100 tablets to a box
  • DPD = N, N-Diethyl-1, 4-phenylene-diamine sulfate
  • Use with existing LaMotte™ chlorine comparators or colorimeters, including the LTC3000W Series Laboratory Turbidity, Chlorine and Color Meter.
Specifications
For Use With (Application) Use existing LaMotte™ chlorine comparators or colorimeters
Quantity 100 Pack
Type Chlorine DPD 3 Instrument Grade Tablet
WARNING: Cancer - www.P65Warnings.ca.gov