Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Chlorine-Free Body Bag with Envelope Zipper
SDP

Catalog No. 22444350 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
94 x 36in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-444-350 94 x 36in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-350 Supplier Mopec Supplier No. BE064

missing translation for 'validMainframeMsg'

Mopec Chlorine-Free Body Bag with Envelope Zipper is impervious to abrasion and tear resistant

  • Material complies with EPA burn restrictions
  • Remains flexible without cracking to -70°F (-57°C) for winter environments or morgue coolers
  • # 8 zipper
  • No handles
  • Static lift tested
  • Heat sealed ends with No Side Seam construction complies with OSHA Reg. 3130

Specifications

Certifications/Compliance Complies with EPA burn restrictions
Closure Type Zip Closure
Color White
Quantity 1
Dimensions (L x W) 94 x 36in.
Capacity (English) 300 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.