Learn More
Brady™ Chlorine Gas High Performance Pipe Marker, 0.375 to 0.5 in.
Shop All Brady Corporation ProductsDescription
- Designed for use on rusty, dirty, wet or rough pipes where pressure sensitive labels aren’t effective.
- Resistant to UV light exposure, high humidity, high temperatures, abrasion, chemical reagents and physical agitation.
- Self-adhesive end strip adheres to the marker's body to ensure a tight, continuous wrap.
- Excellent chemical resistance.
Specifications
Specifications
| Adhesive Type | Non-adhesive |
| Coating | Laminate |
| Color | Black on Orange |
| Display | Text |
| Finish | Gloss |
| Format | Sheet |
| Material | Coiled Plastic (B-689) |
| Product Type | Pipe Marker |
| Sterility | Non-sterile |
| Dimensions (L x W) | 4 x 4 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.