Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorine High Calibration Standard (12 ppm), Reagents

Catalog No. 01335704
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-704 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-704 Supplier Reagents Holdings Llc Supplier No. CC0046001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorine High Calibration Standard (12 ppm), Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Chlorine High Calibration Standard (12 ppm)
Packaging Poly Bottle
CAS 7732-18-5, 1310-73-2
Specific Gravity 1.0042
Physical Form Liquid
Color Colorless
pH 12
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Water Analysis
Quantity 1 L
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.