Learn More
ASI STANDARDS Chlorine in Mineral Oil Check Standard - Low Concentration, 1l
Catalog No. NC3592016
Encompass_Preferred
Chlorine in Mineral Oil manufactured and certified gravimetrically under the guidance of an ISO 9001 quality management system. When applicable, standards are traceable to NIST certified reference materials.Chlorine in Petroleum Oil and Petroleum fuels are manufactured and certified gravimetrically under the guidance of ISO 17034 according to certain methods. Please view our certificate for more information.
Catalog No. NC3592016 Supplier ASI STANDARDS Supplier No. CLMO251L
Chlorine in Mineral Oil manufactured and certified gravimetrically under the guidance of an ISO 9001 quality management system. When applicable, standards are traceable to NIST certified reference materials.Chlorine in Petroleum Oil and Petroleum fuels are manufactured and certified gravimetrically under the guidance of ISO 17034 according to certain methods. Please view our certificate for more information.