Learn More
ASI STANDARDS Chlorine in Water Check Standard 10 mg/kg - 100 mL
Catalog No. NC3592029
Encompass_Preferred
Chlorine in Water manufactured and certified gravimetrically under the guidance of an ISO 9001 quality management system. When applicable, standards are traceable to NIST certified reference materials.ISO 17034 Certified Reference Material
Catalog No. NC3592029 Supplier ASI STANDARDS Supplier No. CLH2O10100
Chlorine in Water manufactured and certified gravimetrically under the guidance of an ISO 9001 quality management system. When applicable, standards are traceable to NIST certified reference materials.ISO 17034 Certified Reference Material