Learn More
ASI STANDARDS Chlorine in Water Check Standard 15 mg/kg - 100 mL
Catalog No. NC3592039
Encompass_Preferred
Chlorine in Water manufactured and certified gravimetrically under the guidance of an ISO 9001 quality management system. When applicable, standards are traceable to NIST certified reference materials.
Catalog No. NC3592039 Supplier ASI STANDARDS Supplier No. CLH2O15100
Chlorine in Water manufactured and certified gravimetrically under the guidance of an ISO 9001 quality management system. When applicable, standards are traceable to NIST certified reference materials.