Learn More
Brim Technologies Sodium Thiosulfate Tablets, 30 mg / 500 count
Catalog No. NC3346457
Encompass_Preferred
Sodium Thiosulfate Tablets, 30 mg / 500 count used to remove residual chlorine from chlorinated water. Safe DChlor T30 Tablets are designed for collection containers from 100-1,500ml
Catalog No. NC3346457 Supplier Brim Technologies Supplier No. T30500
Sodium Thiosulfate Tablets, 30 mg / 500 count used to remove residual chlorine from chlorinated water. Safe DChlor T30 Tablets are designed for collection containers from 100-1,500ml