Learn More
Brim Technologies Sodium Thiosulfate Tablets, 50 mg / 500 count
Catalog No. NC3346459
Encompass_Preferred
Sodium Thiosulfate Tablets, 50 mg / 500 count used to remove residual chlorine from chlorinated water. Safe DChlor T50 Tablets are designed for collection containers from 100-3,000 ml.
Catalog No. NC3346459 Supplier Brim Technologies Supplier No. T50500
Sodium Thiosulfate Tablets, 50 mg / 500 count used to remove residual chlorine from chlorinated water. Safe DChlor T50 Tablets are designed for collection containers from 100-3,000 ml.