Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Lange™ Chlorine Standard Solution, 25-30 mg/L as Cl2, PourRite Ampules (NIST)

Catalog No. 501996391 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Format:
Strip
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
50-199-6391 Strip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-6391 Supplier Hach Lange™ Supplier No. 2630020
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For accuracy checks in chlorine determinations.

For accuracy checks in chlorine determinations. Pack of 20 2-mL ampules. Lot-specific concentration displayed on label.

  • Simple Accuracy Checks
  • Minimize waste
  • Reliable results

  • Simple Accuracy Checks
  • Voluette and PourRite Ampule Standards are designed to work directly with Standard Additions accuracy checks in Hach procedures and photometer software. Use the TenSette Pipet to add precise spikes of standard and check percent recovery, to confirm analytical system performance.
  • Minimize waste
  • Open a fresh standard ampule for each accuracy check. Each ampule contains enough standard for one set of standard additions.
  • Reliable results
  • Sealed ampules prevent accidental contamination or evaporation of standard solutions.

    Specifications

  • Concentration: 25 - 30 mg/L Cl₂
  • Description : Chlorine as Cl₂ (NIST)
  • Parameter: Chlorine
  • Platform: Solution
  • Quantity: 20 Ampules - 2 mL
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 to 2 mL
Format Strip
Type Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.