Learn More
Aqua Solutions Chlorine Standard 100 ppm as Cl₂, NIST Traceable (1L)
SDP
Supplier:  Aqua Solutions 17921L
Encompass_Preferred
Chlorine Standard 100 ppm as Cl₂, NIST Traceable (1L) *This item has a two month expiration date from the date of manufacture.*
Catalog No. NC9202305