Learn More
Grainger Chlorine Test 7 1/2 in L 10 to 200 ppm
Catalog No. 502753498
Encompass_Preferred
The Hydrion Chlorine Test 10 to 200 ppm is a test strip that measures chlorine levels in water. It has a measurement range of 10 to 200 parts per million (ppm) chlorine. The test strips provide fast, accurate chlorine testing and have a shelf life of 1 year if stored properly. They are convenient and easy to use - simply dip the strip in water for 1 second and compare the resulting color to the color chart on the bottle after 30 seconds. This allows you to monitor chlorine levels in pools, spas, drinking water, and more to ensure proper sanitation. The strips come in a bottle of 100.
Catalog No. 50-275-3498 Supplier Grainger Supplier No. 14G845
The Hydrion Chlorine Test 10 to 200 ppm is a test strip that measures chlorine levels in water. It has a measurement range of 10 to 200 parts per million (ppm) chlorine. The test strips provide fast, accurate chlorine testing and have a shelf life of 1 year if stored properly. They are convenient and easy to use - simply dip the strip in water for 1 second and compare the resulting color to the color chart on the bottle after 30 seconds. This allows you to monitor chlorine levels in pools, spas, drinking water, and more to ensure proper sanitation. The strips come in a bottle of 100.