Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Chlorine Test Kit
SDPSCI_ED

Catalog No. s72367 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S72367 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72367 Supplier LaMotte™ Supplier No. 330801
Add to Cart
Add to Cart

Measures free, total and combined chlorine levels

  • Test Method: DPD Tablet and Octa-Slide 2 Comparator
  • Test Range: 0.2 to 3.0 ppm Cl
  • Number of Tests: 50
  • Number of Reagents per test: two
  • DPD = N,N Diethyl-1,4 Phenylenediamine, Sulfate PPM = Parts Per Million, Cl = chemical symbol for Chlorine
  • For testing chemical chlorine levels in drinking water, environmental education, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater

Specifications

Certifications/Compliance NPDWR
For Use With (Application) Drinking water, environmental education, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater
Includes 50 chlorine DPD 1R tablets, 50 chlorine DPD 3R tablets, 2.5 to 5 to 10mL test tubes, with caps, Octa-slide 2 viewer, chlorine octa-slide 2 bar
Quantity 50 Tests
Test Method DPD tablets and Octa-Slide 2 comparator
Type Chemical Chlorine Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.