Learn More
Serim Research Corp Serim GUARDIAN™ Residual Chlorine Test Strips
SDP
Catalog No. NC1766184
Encompass_Preferred
Serim Residual Chlorine Test Strips indicate the concentration of chlorine bleach (sodium hypochlorite) remaining in the solution used to rinse dialysate lines following disinfection of hemodialysis equipment with a chlorine solution. Kit contains 6 bottles of 100 tests
Catalog No. NC1766184 Supplier Serim Research Corp Supplier No. 5100A
Serim Residual Chlorine Test Strips indicate the concentration of chlorine bleach (sodium hypochlorite) remaining in the solution used to rinse dialysate lines following disinfection of hemodialysis equipment with a chlorine solution. Kit contains 6 bottles of 100 tests