Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Chlorine TRACER PockeTester
SDPSCI_ED

Catalog No. S72377 Shop All LaMotte Company Products
Change view
Click to view available options
Data Points:
15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Data Points
S72377 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S72377 Supplier LaMotte™ Supplier No. 1740
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TRACER PockeTester Total Chlorine Kit, Read Total Chlorine from 0.00 to 9.99ppm to 0.01ppm resolution, Extra bold display includes an analog bar graph feature, uses four LR-44 batteries, Readings are not affected by sample color

Reads Total Chlorine from 0.00 to 9.99ppm to 0.01ppm resolution, Extra bold display includes an analog bar graph feature, uses four LR-44 batteries. Readings are not affected by sample color.

Specifications

Type Chlorine Tracer Pocket Tester
Detection Range 0.00 to 9.99 ppm
Display Type LCD
Resolution 0 to 9.99 to 0.01 ppm
Data Points 15
Temperature (English) Operating 23°F to 194°F
For Use With (Application) Industrial water boiling and cooling, laundry and sanitation, water and wastewater
Includes 100 reagent tablets that deliver a precise amount of iodide for a 20mL sample, and four LR-44 batteries. Optional accessories include pH, ORP, and Cl2; sensor probes, and weighted stand with 5 sample cups.
Length (Metric) 18.4 cm
Width (English) 0.93 in.
Width (Metric) 2.8 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.