Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Chlorine ULR Refill Kit
SDP

Catalog No. 13299574
Change view
Click to view available options
Quantity:
30/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-299-574 30/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-574 Supplier CHEMetrics™ by AquaPhoenix Supplier No. R2511
Add to Cart
Add to Cart

Refill kit for K-2511 Chlorine ULR visual test kit used for chlorine analysis in water. Contains 30 self-filling ULR ampoules packaged in trays and a box.

  • Free chlorine oxidizes DDPD, a methyl substituted form of DPD (N,N-diethyl-p-phenylenediamine), to form a purple?colored species in direct proportion to the chlorine concentration.
  • Total chlorine, the sum of free chlorine and combined chlorine (chloramines), is determined by adding potassium iodide (A-2500 solution) to the sample. Chlorine oxidizes the iodide to iodine, and the iodine then oxidizes DDPD to the purple-colored species. Results are expressed in ppm (mg/L) Cl2. The combined chlorine concentration of a sample can be determined by difference between the total and free results

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-2511
No. of Tests 30
Quantity 30/Box
Type Chlorine ULR Refill Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.