Learn More
Cayman Chemical Chlormadinone Acetate, Purity: ≥98%, CAS: 302-22-7, 250 mg
SDP
Catalog No. 502990160
Encompass_Preferred
A synthetic progestin; binds to progesterone, androgen, and glucocorticoid receptors in vitro (Kis = 2.5, 3.8, and 16 nM, respectively, for the human receptors); increases the number of endometrial glands and uterine weight in β-estradiol-primed rabbits at 45 µg/kg per day for five days; reduces testosterone-induced increases in the seminal vesicle weight of castrated male rats at 4.6 and 21.5 mg/kg per day for eight days
Catalog No. 50-299-0160 Supplier Cayman Chemical Supplier No. 29392250 MG
A synthetic progestin; binds to progesterone, androgen, and glucocorticoid receptors in vitro (Kis = 2.5, 3.8, and 16 nM, respectively, for the human receptors); increases the number of endometrial glands and uterine weight in β-estradiol-primed rabbits at 45 µg/kg per day for five days; reduces testosterone-induced increases in the seminal vesicle weight of castrated male rats at 4.6 and 21.5 mg/kg per day for eight days