Learn More
Apexbio Technology LLC CHLORMADINONE-ACETATE-500MG
SDP
Catalog No. 5000570290
Encompass_Preferred
Chlormadinone acetate (CAS No 302-22-7) is a synthetic progestogen commonly used in biomedical research to study hormone-related pathways Originally derived from progesterone it functions primarily by binding to progesterone receptors inhibiting ovulation and exhibiting anti-androgenic properties In vitro it shows bioactivity in the nanomolar to low micromolar range affecting cellular proliferation and gene expression in hormone-sensitive assays It is frequently utilized in cell models including breast cancer and prostate cancer lines as well as in animal studies to explore its effects on reproductive health and hormone-dependent conditions Drug screening applications often seek to understand its modulation of steroid hormone pathways Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70290 Supplier Apexbio Technology LLC Supplier No. B6136500
Chlormadinone acetate (CAS No 302-22-7) is a synthetic progestogen commonly used in biomedical research to study hormone-related pathways Originally derived from progesterone it functions primarily by binding to progesterone receptors inhibiting ovulation and exhibiting anti-androgenic properties In vitro it shows bioactivity in the nanomolar to low micromolar range affecting cellular proliferation and gene expression in hormone-sensitive assays It is frequently utilized in cell models including breast cancer and prostate cancer lines as well as in animal studies to explore its effects on reproductive health and hormone-dependent conditions Drug screening applications often seek to understand its modulation of steroid hormone pathways Experimental concentrations typically depend on the specific research design