Learn More
Medchemexpress LLC HY-A0296 10mg Medchemexpress, Chlormethiazole hydrochloride CAS:6001-74-7 Purity:>98%
SDP
Catalog No. 501970092
Encompass_Preferred
Medchemexpress, HY-A0296 10mg Chlormethiazole HCl CAS:6001-74-7 Clomethiazole hydrochloride is a sedative and anticonvulsant. Clomethiazole hydrochloride is neuroprotective and prevents the degeneration of serotonergic nerve terminals induced by 3,4-methylenedioxymethamphetamine (MDMA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0092 Supplier Medchemexpress LLC Supplier No. HYA029610MG
Medchemexpress, HY-A0296 10mg Chlormethiazole HCl CAS:6001-74-7 Clomethiazole hydrochloride is a sedative and anticonvulsant. Clomethiazole hydrochloride is neuroprotective and prevents the degeneration of serotonergic nerve terminals induced by 3,4-methylenedioxymethamphetamine (MDMA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.