Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlormethiazole Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C3020200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3020200MG 200 mg
C30201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3020200MG Supplier TCI America Supplier No. C3020200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6001-74-7
Molecular Formula C6H9Cl2NS
Molecular Weight (g/mol) 198.11
MDL Number MFCD00673948
InChI Key OFXYKSLKNMTBHK-UHFFFAOYSA-N
Synonym chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1
PubChem CID 19035073
IUPAC Name 5-(2-chloroethyl)-4-methyl-1,3-thiazole hydrochloride
SMILES Cl.CC1=C(CCCl)SC=N1

Specifications

Melting Point 136°C
Color White
Quantity 200 mg
Formula Weight 198.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Chlormethiazole Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.