Learn More
Medchemexpress LLC Chlormezanone | 80-77-3 | MFCD00143951 | 99.8% | 100 MG
SDP
Catalog No. 502082487
Encompass_Preferred
Chlormezanone is a chemical compound that resembles benzodiazepine, acting as an anxiolytic and muscle relaxant. It targets the GABA Receptor and is relevant to neurological and neurodegenerative disease research.
  • Acts as an anxiolytic and muscle relaxant.
  • Resembles benzodiazepine.
  • Targets GABA receptor.
  • Relevant to neurological and neurodegenerative disease research.
Catalog No. 50-208-2487 Supplier Medchemexpress LLC Supplier No. HYB0353100MG
Chlormezanone is a chemical compound that resembles benzodiazepine, acting as an anxiolytic and muscle relaxant. It targets the GABA Receptor and is relevant to neurological and neurodegenerative disease research.
  • Acts as an anxiolytic and muscle relaxant.
  • Resembles benzodiazepine.
  • Targets GABA receptor.
  • Relevant to neurological and neurodegenerative disease research.