Learn More
Medchemexpress LLC Chlormezanone | 80-77-3 | 99.8% | 200 MG
SDP
Catalog No. 5000407210
Encompass_Preferred
Chlormezanone resembles benzodiazepine, and its action is similar to benzodiazepine-type agents. It is used as an anxiolytic and a muscle relaxant.
  • Used as an anxiolytic and a muscle relaxant.
  • Resembles benzodiazepine and acts similarly to benzodiazepine-type agents.
  • Available in solid form (white to off-white).
  • For research use only.
Catalog No. 50-004-07210 Supplier Medchemexpress LLC Supplier No. HYB0353200MG
Chlormezanone resembles benzodiazepine, and its action is similar to benzodiazepine-type agents. It is used as an anxiolytic and a muscle relaxant.
  • Used as an anxiolytic and a muscle relaxant.
  • Resembles benzodiazepine and acts similarly to benzodiazepine-type agents.
  • Available in solid form (white to off-white).
  • For research use only.