Learn More
Cayman Chemical Chlormezanone, Purity: ≥98%, CAS: 80-77-3, 500 mg
SDP
Catalog No. 502935901
Encompass_Preferred
A muscle relaxant; induces paralysis in mice, guinea pigs, dogs, cats, and monkeys (EC50s = 133, <200, 100-250, 50, and 50 mg/kg, respectively); increases ataxia induced by pentobarbital and alcohol in mice; blocks the crossed extensor reflex of the spine without affecting the knee-jerk reflex in cats; induces head drop in rabbits
Catalog No. 50-293-5901 Supplier Cayman Chemical Supplier No. 25716500 MG
A muscle relaxant; induces paralysis in mice, guinea pigs, dogs, cats, and monkeys (EC50s = 133, <200, 100-250, 50, and 50 mg/kg, respectively); increases ataxia induced by pentobarbital and alcohol in mice; blocks the crossed extensor reflex of the spine without affecting the knee-jerk reflex in cats; induces head drop in rabbits