Learn More
Medchemexpress LLC Chlormidazole (hydrochloride) | 74298-63-8 | 99.5% | 25 MG
SDP
Catalog No. 5000413925
Encompass_Preferred
Chlormidazole hydrochloride is an antifungal agent with inhibitory activity against many fungi and some gram-positive cocci. It can be applied in fungal and bacterial infections of nails and skin, including interdigital and periungual mycoses.
  • Inhibits fungal growth.
  • Inhibits some gram-positive cocci.
  • Suitable for research use only.
Catalog No. 50-004-13925 Supplier Medchemexpress LLC Supplier No. 25MGHYB1144A
Chlormidazole hydrochloride is an antifungal agent with inhibitory activity against many fungi and some gram-positive cocci. It can be applied in fungal and bacterial infections of nails and skin, including interdigital and periungual mycoses.
  • Inhibits fungal growth.
  • Inhibits some gram-positive cocci.
  • Suitable for research use only.