Learn More
AdipoGen Chlornitrofen
SDP
Supplier:  AdipoGen CDXC0233G001
Encompass_Preferred
Chemical. CAS 1836-77-7. Formula C12H6Cl3NO3. MW 318.54. Synthetic. Herbicide that was used to control annual weeds in paddy fields. Protoporphyrinogen oxidase inhibitor. Compound can be used as analytical reference material.
Catalog No. 50-256-1418