Learn More
Strem, An Ascensus Company CAS# 578743-87-0. 2g. Chloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]copper(I), 98%. MFCD09264276
SDP
Catalog No. 5090111325
Encompass
CAS# 578743-87-0. 2g. Chloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]copper(I), 98%. MFCD09264276. Molecular Weight: 487.59. Molecular Formula: C27H36ClCuN2. Color/form: white pwdr. Strem# 29-4050. http://www.strem.com/catalog/v/29-4050/
Catalog No. 50-901-11325 Supplier Strem, An Ascensus Company Supplier No. 2940502G
CAS# 578743-87-0. 2g. Chloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]copper(I), 98%. MFCD09264276. Molecular Weight: 487.59. Molecular Formula: C27H36ClCuN2. Color/form: white pwdr. Strem# 29-4050. http://www.strem.com/catalog/v/29-4050/