Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I) 97.0+%, TCI America™
SDP

Catalog No. C2422200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2422200MG 200 mg
C24221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2422200MG Supplier TCI America Supplier No. C2422200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 873779-78-3
Molecular Formula C21H24ClCuN2
Synonym [1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]chlorocopper(I)

Specifications

Melting Point 277°C
Quantity 200 mg
Formula Weight 403.43
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I)
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.